Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OBFC2B Antibody (OTI1E11), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273140PE
Description
OBFC2B Monoclonal specifically detects OBFC2B in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
OBFC2B | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
NABP2 | |
Full length recombinant protein of human OBFC2B (NP_076973) produced in HEK293T cells. | |
0.1 mL | |
79035.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI1E11 | |
Flow Cytometry | |
LP3587, MGC2731, oligonucleotide/oligosaccharide-binding fold containing 2B, Oligonucleotide/oligosaccharide-binding fold-containing protein 2B, Sensor of single-strand DNA complex subunit B1, Sensor of ssDNA subunit B1, Single-stranded DNA-binding protein 1, SOSS complex subunit B1, SOSS-B1, SSB1hSSB1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction