Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OBP/Aphrodisin Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12488R

Description
The protein encoded by this gene is a member of kinesin-like protein family. This family of proteins are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. The C-terminal half of this protein has been shown to bind DNA. Studies with the Xenopus homolog suggests its essential role in metaphase chromosome alignment and maintenance.Specifications
OBP/Aphrodisin | |
Polyclonal | |
Unconjugated | |
KIF22 | |
A-328A3.2; AU021460; C81217; KID; Kif22; Kif22a; kinesin family member 22; kinesin family member 22A; kinesin-like 4; Kinesin-like DNA-binding protein; kinesin-like DNA-binding protein pseudogene; Kinesin-like protein 4; Kinesin-like protein KIF22; KNSL4; OBP; OBP-1; OBP-2; origin of plasmid DNA replication-binding protein; oriP binding protein; SEMDJL2 | |
Rabbit | |
Protein A | |
RUO | |
3835 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q14807 | |
KIF22 | |
KLH conjugated synthetic peptide derived from rat OBP/Aphrodisin. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction