Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    OBP2B Antibody (002), HRP, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290310H
Description
OBP2B Monoclonal specifically detects OBP2B in Human samples. It is validated for ELISA.Specifications
| OBP2B | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 29989 | |
| Store at 4°C in the dark. | |
| IgG | 
| ELISA | |
| 2 | |
| ELISA | |
| hOBPIIb, MGC119022, OBPIIb, odorant binding protein 2B, odorant-binding protein 2b, odorant-binding protein IIb, UNQ653/PRO1283 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human OBP2B (NP_055396.1; Met1-His170). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction