Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OBP2B Antibody (002), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290310PE
Description
OBP2B Monoclonal antibody specifically detects OBP2B in Human samples. It is validated for ELISASpecifications
OBP2B | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
29989 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
002 | |
ELISA | |
hOBPIIb, MGC119022, OBPIIb, odorant binding protein 2B, odorant-binding protein 2b, odorant-binding protein IIb, UNQ653/PRO1283 | |
Rabbit immunized with purified, recombinant Human OBP2B (Accession#: NP_055396.1, Met1-His170) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction