Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OBP2B Rabbit anti-Human, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290310100UL
Description
OBP2B Monoclonal specifically detects OBP2B in Human samples. It is validated for ELISA.Specifications
OBP2B | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
29989 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
ELISA | |
2 | |
ELISA 1:5000-1:10000 | |
hOBPIIb, MGC119022, OBPIIb, odorant binding protein 2B, odorant-binding protein 2b, odorant-binding protein IIb, UNQ653/PRO1283 | |
Recombinant Human OBP2B Protein | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction