Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Obscurin Mouse anti-Human, Mouse, Rat, Clone: 5H10A, MilliporeSigma™

Catalog No. MABT126MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT126MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT126MI Supplier MilliporeSigma Supplier No. MABT126
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Obscurin, clone 5H10A Antibody and Western Blotting for the detection of obscurin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Obscurin
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 5H10A
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols OBSCN,KIAA1556,KIAA1639
Host Species Mouse
Immunogen GST-tagged full-length human recombinant Obscurin.
Purification Method Protein G Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001092093
Test Specificity Clone 5H10A detects obscurin in heart and skeletal muscle.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.