Learn More
Eisco™ Obsidian Specimen
SCI_ED
Catalog No. S24763 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24763 12/Pk. 2 to 3 cm
S24762 1 Ea. 2 to 3 cm
S26587 10/Pk. 2 to 3 cm
S26630 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24763 Supplier Eisco™ Supplier No. ESNG0046PK12
Add to Cart
Add to Cart
Obsidian Specimen, also known as volcanic glass, forms from lava that cools almost immediately upon eruption.
  • Volcanic glass formed through nearly instantaneous quenching of lava
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Obsidian shares many physical properties with its major chemical component, quartz, despite looking dissimilar
  • Despite being dark in color, obsidian is made up of microcrystalline quartz, demonstrated by sharing hardness and conchoidal fracture often exhibited by quartz
  • For use in classroom identification labs and exercises
Specifications
Product Type Igneous Rock
For Use With (Application) Classroom identification labs and exercises
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Obsidian