Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OC90 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571564
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571564 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571564 Supplier Invitrogen™ Supplier No. PA571564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OC90 Polyclonal Antibody for Western Blot, Flow

Major protein of the otoconia, a calcium carbonate structure in the saccule and utricle of the ear. Together with OTOL1, acts as a scaffold for otoconia biomineralization: sequesters calcium and forms interconnecting fibrils between otoconia that are incorporated into the calcium crystal structure. Together with OTOL1, modulates calcite crystal morphology and growth kinetics. It is unlikely that this protein has phospholipase A2 activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen OC90
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OC90
Gene Accession No. Q02509
Gene Alias OC90; otoconin 90; otoconin-90; Phospholipase A2 homolog; PLA2L
Gene Symbols OC90
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 223-252 amino acids from the Central region of human OC90.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729330
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.