Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OCA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598335
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598335 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598335 Supplier Invitrogen™ Supplier No. PA598335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OCA2 Polyclonal Antibody for Western Blot, ELISA

This gene encodes the human homologue of the mouse p (pink-eyed dilution) gene. The encoded protein is believed to be an integral membrane protein involved in small molecule transport, specifically tyrosine - a precursor of melanin. Mutations in this gene result in type 2 oculocutaneous albinism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCA2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene OCA2
Gene Accession No. Q04671
Gene Alias BEY; BEY1; BEY2; BOCA; D15S12; D7H15S12; D7Icr28RN; D7Nic1; EYCL; EYCL2; EYCL3; eye color 2 (central brown); eye color 3 (brown); hair color 3 (brown); HCL3; Melanocyte-specific transporter protein; Oca2; OCA2 melanosomal transmembrane protein; oculocutaneous albinism II; oculocutaneous albinism II (pink-eye dilution homolog, mouse); p; P protein; PED; pink-eyed dilution protein; Pink-eyed dilution protein homolog; P-protein; SHEP1; total brown iris pigmentation
Gene Symbols OCA2
Host Species Rabbit
Immunogen Recombinant Human P protein (2-136AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4948
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.