Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OCAM/NCAM2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297119AF594
Description
OCAM/NCAM2 Polyclonal specifically detects OCAM/NCAM2 in Human samples. It is validated for ELISA.Specifications
OCAM/NCAM2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 594 | |
N-CAM-2, NCAM-2, NCAM21MGC51008, neural cell adhesion molecule 2, neural cell adhesion molecule 2 precursor10 | |
Produced in rabbits immunized with purified, recombinant OCAM/NCAM2 (NP_004531.2; Met1-Asn697). | |
0.1 mL | |
Growth and Development, Membrane Vesicle Markers, Neuronal Cell Markers | |
4685 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction