Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564212
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564212 Supplier Invitrogen™ Supplier No. PA564212
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGCGNSTATS AGAGQGPAGA AKDVTEESVT EDDKRRNYGG VYVGLPSEAV NMVSSQTKTV RK Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 52%.

OCC-1 (Overexpressed in colon carcinoma-1) was initially identified as a novel human cDNA overexpressed in a colon carcinoma cell line. Elevated levels of OCC-1 mRNA were observed in three of eight colon carcinomas, suggesting that OCC-1 may be a hallmark of only a subset of colon carcinomas. It is highly expressed in skeletal muscle, kidney, placenta, and pancreas tissues, with low to no expression in heart, brain, lung and liver tissues. At least two different OCC-1 mRNA species are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C12orf75
Gene Accession No. Q8TAD7
Gene Alias adipogenesis down-regulated 3; AGD3; C12orf75; chromosome 12 open reading frame 75; OCC1; OCC-1; Overexpressed in colon carcinoma 1 protein; overexpressed in colon carcinoma-1; putative overexpressed in colon carcinoma-1 protein variant C
Gene Symbols C12orf75, OCC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OCC1. Recombinant protein control fragment (Product #RP-103594).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387882
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.