Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144307
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144307 Supplier Invitrogen™ Supplier No. PA5144307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OCC1 Polyclonal Antibody for IHC (P), ELISA

OCC-1 (Overexpressed in colon carcinoma-1) was initially identified as a novel human cDNA overexpressed in a colon carcinoma cell line. Elevated levels of OCC-1 mRNA were observed in three of eight colon carcinomas, suggesting that OCC-1 may be a hallmark of only a subset of colon carcinomas. It is highly expressed in skeletal muscle, kidney, placenta, and pancreas tissues, with low to no expression in heart, brain, lung and liver tissues. At least two different OCC-1 mRNA species are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCC1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3.055 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene C12orf75
Gene Accession No. Q8TAD7
Gene Alias adipogenesis down-regulated 3; AGD3; C12orf75; chromosome 12 open reading frame 75; OCC1; OCC-1; Overexpressed in colon carcinoma 1 protein; overexpressed in colon carcinoma-1; putative overexpressed in colon carcinoma-1 protein variant C
Gene Symbols C12orf75, OCC1
Host Species Rabbit
Immunogen Recombinant Human Overexpressed in colon carcinoma 1 protein (1-63AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387882
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.