Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Occludin Monoclonal Antibody (OC-3F10), Abbomax

Catalog No. 502204415
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4415 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4415 Supplier Abbomax Inc Supplier No. 606110
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Breast tissue Cellular location: Membrane, cell junction.

Occludin Monoclonal antibody specifically detects Occludin in Bacteria, Chicken, Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Occludin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OC-3F10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene OCLN
Gene Accession No. Q16625, Q6P6T5, Q91049
Gene Alias AI503564; BLCPMG; Occludin; occludin 1B; Ocl; Ocln; phosphatase 1, regulatory subunit 115; PPP1R115; tight junction protein occludin; tight junction structural protein
Host Species Mouse
Immunogen A recombinant protein consisting of the C-terminal region (150aa) of human Occludin
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100506658, 396026, 83497
Target Species Bacteria, Chicken, Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.