Learn More
Invitrogen™ Occludin Monoclonal Antibody (OC-3F10), Alexa Fluor™ 594

Description
Reactivity of this antibody with the occludin protein has been confirmed by immunofluorescence and Western blotting (un-conjugated). Tissues/lysates tested: T84 cell line (human intestinal epithelium), MDCK cells (canine kidney), Caco-2 cells (human colon adenocarcinoma), MTE7B (Mouse) and rat liver This antibody reacts specifically with mammalian occludin.
Specifications
Specifications
| Antigen | Occludin |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OC-3F10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Alexa Fluor 594 |
| Formulation | PBS with 4mg/mL BSA and 0.1% sodium azide; pH 7.4 |
| Gene | OCLN |
| Gene Accession No. | Q16625, Q28269, Q61146, Q6P6T5 |
| Gene Alias | AI503564; BLCPMG; Occludin; occludin 1B; Ocl; Ocln; phosphatase 1, regulatory subunit 115; PPP1R115; tight junction protein occludin; tight junction structural protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.