Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Occludin Mouse anti-Human, Clone: OCLN/2183, Novus Biologicals™

Mouse Monoclonal Antibody
$280.00
Specifications
Antigen | Occludin |
---|---|
Clone | OCLN/2183 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
Occludin Monoclonal specifically detects Occludin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
Occludin | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
BLCPMG, occludin | |
Recombinant human Occludin fragment around aa 282-415 (Exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
OCLN/2183 | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
100506658 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title