Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Occludin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1D3C4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Occludin |
---|---|
Clone | 1D3C4 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6807
![]() ![]() |
Proteintech Group Inc
663781IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes an integral membrane protein that is required for cytokine-induced regulation of the tight junction paracellular permeability barrier. Mutations in this gene are thought to be a cause of band-like calcification with simplified gyration and polymicrogyria (BLC-PMG), an autosomal recessive neurologic disorder that is also known as pseudo-TORCH syndrome. Alternative splicing results in multiple transcript variants. A related pseudogene is present 1.5 Mb downstream on the q arm of chromosome 5.Specifications
Occludin | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Occludin, OCLN | |
Ocln | |
IgG1 | |
Protein G | |
Antibody |
1D3C4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
Q16625, Q61146, Q6P6T5 | |
100506658, 18260, 397236, 83497 | |
Occludin Fusion Protein Ag4057 | |
Primary | |
-20°C | |
OCLN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title