Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Occludin Recombinant Mouse Monoclonal Antibody (OC-3F10), Alexa Fluor™ Plus 594, Invitrogen™
Mouse Recombinant Monoclonal Antibody
$123.65 - $323.00
Specifications
Antigen | Occludin |
---|---|
Clone | OC-3F10 |
Concentration | 1.0 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
74-000-6MP594
![]() |
Invitrogen™
740006MP594 |
50 μL |
Each for $323.00
|
|
|||||
50-253-9566
![]() |
Invitrogen™
740006MP59420UG |
20 μL |
Each
for $123.65
Save 25% Reg : |
|
|||||
Description
Occludin is a 65 kDa protein that can exist in a variety of phosphorylated forms, ranging up to approximately 82 kDa. Occludin is thought to be involved in regulating both the localization and the function of occludin. Polyunsaturated fatty acids are known to up-regulate occludin expression, increasing the transendothelial cell resistance and reducing the cellular permeability to large molecules. The level of occludin varies greatly depending on tissue; in brain tissue, occludin is highly and continuously expressed at cell-cell contact sites, whereas non-neural tissues show lower expression and discontinuous distribution. Overall structural features of the occludin protein are highly conserved in all the species examined. Under-expression of tight junction proteins, including occludin, is a key molecular abnormality responsible for the increased permeability of tumor endothelial tight junctions, which contributes to brain tumor edemas.Specifications
Occludin | |
1.0 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5 | |
AI503564; BLCPMG; Occludin; occludin 1B; Ocl; Ocln; phosphatase 1, regulatory subunit 115; PPP1R115; tight junction protein occludin; tight junction structural protein | |
OCLN | |
IgG1 κ | |
Protein A/G | |
OCLN |
OC-3F10 | |
Flow Cytometry, Immunocytochemistry | |
Alexa Fluor Plus 594 | |
Mouse | |
Human | |
Q16625 | |
100506658 | |
GST fusion protein consisting of the C-terminal region (~150aa) of human occludin. | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title