Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Occludin Recombinant Superclonal™ Antibody (6HCLC)
Occludin Recombinant Polyclonal Antibody (6HCLC)
Supplier: Thermo Scientific 710192
Description
Occludin Recombinant Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
Occludin | |
Recombinant Superclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q16625, Q61146 | |
OCLN | |
Peptide corresponding to amino acids 387-403 of human occuludin. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
6HCLC | |
Unconjugated | |
OCLN | |
AI503564; BLCPMG; Occludin; occludin 1B; Ocl; Ocln; phosphatase 1, regulatory subunit 115; PPP1R115; tight junction protein occludin; tight junction structural protein | |
Rabbit | |
Protein A | |
RUO | |
100506658, 18260 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction