Learn More
Description
Please see the protocol for further information on protein/antibody labeling and conjugation.
Specifications
Specifications
| Chemical Name or Material | 3-Carboxy-6,8-difluoro-7-hydroxycoumaryl succinimidyl ester |
| CAS | 215868-33-0 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | Standard Fluorescein, Coumarin and Rhodamine-based Dyes |
| Molecular Formula | C14H7F2NO7 |
| Formula Weight | 339.2 |
| Product Type | Blue Fluorescent Dye |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

