Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ochratoxin A, 99+%

Catalog No. ac227400050
Click to view available options
Quantity:
5 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Ochratoxin A, CAS # 303-47-9, is a mycotoxin produced by certain fungi. | CAS: 303-47-9 | C20H18ClNO6 | 403.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Ochratoxin A is a mycotoxin of fungal origin, particularly from Aspergillus and Penicillium species, commonly found in spoiled batches of grains, beans, and nuts
• Ochratoxin A is a known mutagen and potential carcinogen; suspected to affect fetal development and promote the development of urinary tract tumors
Applications
• Ochratoxin A can be used as a calcium channel blocker in cells
• May be useful for developmental research studies
• Intended for research applications only

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-47-9
Molecular Formula C20H18ClNO6
Molecular Weight (g/mol) 403.82
MDL Number MFCD00078079
InChI Key RWQKHEORZBHNRI-BMIGLBTASA-N
Synonym ochratoxin a, antibiotic 9663, phenylalanine-ochratoxin a, 3r14s-ochratoxin a, ochratoxin a-bsa conjugate from aspergillus ochraceus, --n-5-chloro-8-hydroxy-3-methyl-1-oxo-7-isochromanyl carbonyl-3-phenylalanine, n-3r-5-chloro-8-hydroxy-3-methyl-1-oxo-7-isochromanyl carbonyl-3-phenyl-l-alanine, unii-1779sx6luy, l-phenylalanine, n-5-chloro-3,4-dihydro-8-hydroxy-3-methyl-1-oxo-1h-2-benzopyran-7-yl carbonyl-, r, ccris 2382
PubChem CID 442530
ChEBI CHEBI:7719
IUPAC Name (2S)-2-[[(3R)-5-chloro-8-hydroxy-3-methyl-1-oxo-3,4-dihydroisochromene-7-carbonyl]amino]-3-phenylpropanoic acid
SMILES CC1CC2=C(C=C(C(=C2C(=O)O1)O)C(=O)NC(CC3=CC=CC=C3)C(=O)O)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 169.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 mg
Beilstein 18, V,9, 56
Merck Index 15, 6829
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform, diethyl ether, benzene,, ethanol, methanol
Formula Weight 403.82
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Ochratoxin A, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
Fatal if swallowed.
Causes serious eye irritation.
Suspected of damaging the unborn child.
Fatal if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
EINECSNumber 206-143-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.