Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ochratoxin-A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Ochratoxin-A |
---|---|
Concentration | 5.0 mg/mL |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017510
![]() |
Novus Biologicals
NB100627930.1ML |
0.1 mL |
Each for $181.90
|
|
|||||
NB10062793
![]() |
Novus Biologicals
NB10062793 |
0.5 mL |
Each for $487.50
|
|
|||||
Description
Ochratoxin-A Polyclonal specifically detects Ochratoxin-A in Fungi samples. It is validated for ELISA.Specifications
Ochratoxin-A | |
ELISA 1:100-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.09% Sodium Azide | |
Ochratoxin A conjugated to BSA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5.0 mg/mL | |
ELISA | |
Unconjugated | |
Rabbit | |
Fungi | |
OA, Ochratoxin A non-ribosomal peptide synthetase, Ochratoxin A polyketide synthase, OchratoxinA, OTA | |
IgG | |
Protein G purified | |
NB100-62793 is specific for Ochratoxin A. Is shows 1% reactivity with Ochratoxin B 1% and <0.1% reactivity with BSA. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title