Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OCIAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100655 Supplier Invitrogen™ Supplier No. PA5100655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OCIAD1.

Maintains stem cell potency. Increases STAT3 phosphorylation and controls ERK phosphorylation. May act as a scaffold, increasing STAT3 recruitment onto endosomes. Involved in integrin-mediated cancer cell adhesion and colony formation in ovarian cancer (PubMed:20515946). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCIAD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ociad1
Gene Accession No. Q5XIG4, Q9CRD0, Q9NX40
Gene Alias 6030432N09Rik; AI481327; ASRIJ; AW557942; B230209J16Rik; BB021357; Emi2; expressed during mesenchymal induction 2; FLJ20455; Imi2; induced during mesenchymal induction 2; LOC100723738; MGC111072; OCIA; OCIA domain containing 1; OCIA domain-containing protein 1; Ociad1; OCIAD1 protein; Ovarian cancer immunoreactive antigen domain containing 1; Ovarian carcinoma immunoreactive antigen; TPA018
Gene Symbols Ociad1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OCIAD1(Accession Q9NX40), corresponding to amino acid residues I84-Y134.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289590, 54940, 68095
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.