Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OCIAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520834
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520834 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520834 Supplier Invitrogen™ Supplier No. PA520834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is 293 cell lysate. PA5-20834 can be used with blocking peptide PEP-0948.

Maintains stem cell potency. Increases STAT3 phosphorylation and controls ERK phosphorylation. May act as a scaffold, increasing STAT3 recruitment onto endosomes. Involved in integrin-mediated cancer cell adhesion and colony formation in ovarian cancer (PubMed:20515946). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCIAD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Ociad1
Gene Accession No. Q9NX40
Gene Alias 6030432N09Rik; AI481327; ASRIJ; AW557942; B230209J16Rik; BB021357; Emi2; expressed during mesenchymal induction 2; FLJ20455; Imi2; induced during mesenchymal induction 2; LOC100723738; MGC111072; OCIA; OCIA domain containing 1; OCIA domain-containing protein 1; Ociad1; OCIAD1 protein; Ovarian cancer immunoreactive antigen domain containing 1; Ovarian carcinoma immunoreactive antigen; TPA018
Gene Symbols Ociad1
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the carboxy terminus of human OCIAD1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54940
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.