Learn More
Invitrogen™ OCIAD2 Polyclonal Antibody

Description
A suggested positive control is SK-N-SH cell lysate. PA5-20836 can be used with blocking peptide PEP-0950.
Specifications
Specifications
| Antigen | OCIAD2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | OCIAD2 |
| Gene Accession No. | Q56VL3, Q9D8W7 |
| Gene Alias | 1810027I20Rik; OCIA domain containing 2; OCIA domain-containing protein 2; OCIAD2; Ovarian carcinoma immunoreactive antigen-like protein |
| Gene Symbols | OCIAD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.