Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCIAD2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987331
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7331 Supplier Bioss Supplier No. BS6557R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OCIAD2 was identified by its sequence similarity with OCIAD1, and together OCIAD1 and OCIAD2 form the OCIA domain family. OCIAD2 mRNA was found to be expressed at higher levels in invasive adenocarcinoma mixed subtype with bronchioloalveolar carcinoma component (BAC) of the lung. Loss of OCIAD2 expression was significantly correlated with lymphatic invasion, blood vessel invasion, and lymph node metastasis, indicating that OCIAD2 may play a role in cell adhesion and prevention of cell migration. While the function of OCIAD2 is still unknown, its expression in adenocarcinoma with BAC component is significantly associated with a favorable prognosis and may serve as a marker for selecting tumors that are treatable by limited surgery.

Specifications

Antigen OCIAD2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene OCIAD2
Gene Accession No. Q56VL3
Gene Alias 1810027I20Rik; OCIA domain containing 2; OCIA domain-containing protein 2; OCIAD2; Ovarian carcinoma immunoreactive antigen-like protein
Gene Symbols OCIAD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human OCIAD2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132299
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.