Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OCIL/CLEC2d Antibody (166C1309), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056553AF700
Description
OCIL/CLEC2d Monoclonal specifically detects OCIL/CLEC2d in Mouse samples. It is validated for Western Blot.Specifications
OCIL/CLEC2d | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CLEC2D | |
This antibody was developed by immunizing mice with a synthetic peptide containing amino acids 2-18 (CVTKASLPMLSPTGSPQ) of mouse OCIL, GenBank no. gb AAL37198.1. | |
0.1 mL | |
Immunology | |
29121 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
166C1309 | |
Western Blot | |
CLAXLLT-1, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1Osteoclast inhibitory lectin, OCILmember D | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction