Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OCRL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552938 Supplier Invitrogen™ Supplier No. PA552938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DYFLTISGNY LPSCFGTSLE ALCRMKRPIR EVPVTKLIDL EEDSFLEKEK SLLQMVPLDE GASERPLQVP KEIWLLVDHL FKYACHQEDL FQTPGMQEEL QQIIDCLDTS IPETIPGSNH SVAEALLIFL EALPEPVICY ELY Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.

The inositol polyphosphate 5-phosphatases selectively remove the phosphate from the 5-position of various phosphatidylinositols, which generate second messengers in response to extracellular signals. OCRL1 is a type II 5-phosphatase that is mutated in the oculocerebrorenal syndrome of Lowe (OCRL). OCRL is a rare X-linked disorder that is characterized in part by congenital cataracts, mental retardation, muscular hypotonia and renal tubular dysfunction. OCRL1 has a high affinity for phosphatidylinositol 4,5-bisphosphate as well as inositol 1,4,5-trisphosphate and inositol 1,3,4,5-tetrakisphosphate as substrates. OCRL1 is localized to the Golgi complex and is thought to be part of the trans-Golgi network (TGN), which suggests that OCRL1 plays a role in protein sorting and trafficking within the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCRL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OCRL
Gene Accession No. Q01968
Gene Alias 9530014D17Rik; BB143339; Inositol polyphosphate 5-phosphatase OCRL; Inositol polyphosphate 5-phosphatase OCRL-1; INPP5F; LOCR; Lowe oculocerebrorenal syndrome protein; NPHL2; Ocrl; OCRL, inositol polyphosphate-5-phosphatase; OCRL1; OCRL-1; oculocerebrorenal syndrome of Lowe; Phosphatidylinositol 3,4,5-triphosphate 5-phosphatase; phosphatidylinositol polyphosphate 5-phosphatase
Gene Symbols OCRL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OCRL. Recombinant protein control fragment (Product #RP-92194).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4952
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.