Learn More
OCT-1 Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 601440

Description
Positive control: Jurkat Cell Lysate, Human endometrium Cellular location: Nucleus.
2022-10-01 00:00:00 Polyclonal antibody specifically detects 2022-10-01 00:00:00 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
2022-10-01 00:00:00 | |
Polyclonal | |
Unconjugated | |
POU2F1 | |
2810482H01Rik; NF-A1; OCT1; oct-1; oct-1B; octamer binding protein 1; octamer-binding protein 1; Octamer-binding transcription factor 1; Octamer-binding transcription factor-1; Otf1; OTF-1; POU class 2 homeobox 1; POU domain, class 2, transcription factor 1; Pou2f1; ubiquitous transcription factor | |
A synthetic peptide corresponding to C-terminus of OCT-1 protein | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P14859, P25425 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18986, 5451 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.