Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Oct-2 Recombinant Rabbit Monoclonal Antibody (JE58-05)
GREENER_CHOICE

Catalog No. PIMA542519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542519 Supplier Invitrogen™ Supplier No. MA542519
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: Daudi cell lysates, human tonsil tissue, Daudi. Subcellular Location: Nucleus, Cytoplasm.

OCT-2 is a transcription factor belonging to the POU homeo-domain family that binds to the Ig gene octamer sites regulating B cell specific genes. OCT-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. OCT-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oct-2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE58-05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene POU2F2
Gene Accession No. P09086, Q00196
Gene Alias 2-Oct; homeobox protein; lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; OCT2; Oct-2; Oct2a; Oct2b; octamer-binding protein 2; Octamer-binding transcription factor 2; OTF2; OTF-2; POU class 2 homeobox 2; POU domain, class 2, transcription factor 2; Pou2f2
Gene Symbols POU2F2
Host Species Rabbit
Immunogen Recombinant full length protein of human Oct-2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117058, 18987, 5452
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.