Learn More
Oct-4 Rabbit anti-Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304591A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Human, Bovine, Pig and Monkey
Oct1 or POU2F1 (Pou domain, class 2, transcription factor 1) is a transcription factor that binds to the octamer motif (5′-ATTTGCAT-3′) and activates the promoters of the genes for some small nuclear RNAs (snRNA) and of genes such as those for histone H2B and immunoglobulins. It modulates transcription transactivation by NR3C1, AR, and PGR and interacts with NR3C1, AR, and PGR. Oct1 is ubiquitous and belongs to the POU transcription factor family class 2 subfamily. Oct1 contains 1 homeobox domain.Specifications
Oct-4 | |
Polyclonal | |
Unconjugated | |
Pou5f1 | |
Octamer-binding transcription factor 3 antibody, Oct-3, OCT3, POU domain class 5 transcription factor 1 antibody, Oct-4, OTF3 antibody. | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18999 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P20263 | |
Pou5f1 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.