Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oct-4A Antibody (653108), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB17591Z
Description
Oct-4A Monoclonal antibody specifically detects Oct-4A in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
Oct-4A | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5460 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
653108 | |
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
OCT3;Oct-3;OCT4;Oct-4;OCT4;Oct3/4;OTF3;OTF-3;OTF4;POU domain protein;POU domain, class 5, transcription factor 1;POU5F1 | |
E. coli-derived recombinant human Oct-4A, Met1-Glu135, Accession # NP_002692 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction