Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oct-4B Antibody (OTI7H1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273141B
Description
Oct-4B Monoclonal specifically detects Oct-4B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Oct-4B | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI7H1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
POU5F1 | |
Full length recombinant protein of human Oct-4B (NP_002692) produced in HEK293T cells. | |
0.1 mL | |
5460.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction