Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OCT1 (POU2F1) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523392
Description
OCT1 (POU2F1) Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
OCT1 (POU2F1) is a ubiquitously expressed transcription factor that binds to the octamer motif (5'-ATTTGCAT-3') and activates the promoters of the genes for some small nuclear RNAs (snRNA) and of genes such as those for histone H2B and immunoglobulins. It modulates transcription transactivation by NR3C1, AR and PGR. In case of human herpes simplex virus (HSV) infection, POU2F1 forms a multiprotein-DNA complex with the viral transactivator protein VP16 and HCFC1 thereby enabling the transcription of the viral immediate early genes.
Specifications
OCT1 (POU2F1) | |
Polyclonal | |
Unconjugated | |
POU2F1 | |
2810482H01Rik; NF-A1; OCT1; oct-1; oct-1B; octamer binding protein 1; octamer-binding protein 1; Octamer-binding transcription factor 1; Octamer-binding transcription factor-1; Otf1; OTF-1; POU class 2 homeobox 1; POU domain, class 2, transcription factor 1; Pou2f1; ubiquitous transcription factor | |
Rabbit | |
Protein A | |
RUO | |
100051363, 490360, 5451 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
P14859 | |
POU2F1 | |
A portion of amino acids 1-50 of human OCT-1. | |
100 μg | |
Primary | |
Human, Canine, Monkey, Horse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction