Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OCT2 Antibody (OCT2/7073R), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314146AF532
This item is not returnable.
View return policy
Description
OCT2 Monoclonal antibody specifically detects OCT2 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
OCT2. | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OCT2/7073R | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
homeobox protein, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2, OCT2Oct-2, Octamer-binding protein 2, Octamer-binding transcription factor 2, OTF-2, OTF2POU domain class 2, transcription factor 2, POU class 2 homeobox 2, POU domain, class 2, transcription factor 2 | |
Recombinant fragment of human OCT2 protein (around aa 112-297) (exact sequence is proprietary) (Uniprot: P09086) | |
0.1 mL | |
Cancer | |
5452 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction