Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OCT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104461 Supplier Invitrogen™ Supplier No. PA5104461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OCT2.

OCT-2 is a transcription factor belonging to the POU homeo-domain family that binds to the Ig gene octamer sites regulating B cell specific genes. OCT-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. OCT-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCT2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene POU2F2
Gene Accession No. P09086, Q00196
Gene Alias 2-Oct; homeobox protein; lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; OCT2; Oct-2; Oct2a; Oct2b; octamer-binding protein 2; Octamer-binding transcription factor 2; OTF2; OTF-2; POU class 2 homeobox 2; POU domain, class 2, transcription factor 2; Pou2f2
Gene Symbols POU2F2
Host Species Rabbit
Immunogen A synthesized peptide derived from human POU2F2(Accession P09086), corresponding to amino acid residues M1-E34.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117058, 18987, 5452
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.