Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501726669 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6669 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6669 Supplier Proteintech Group Inc Supplier No. 108672AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Oct2 is a transcription factor that specifically binds to the octamer motif (5′-ATTTGCAT-3′). Oct2 regulates transcription in a number of tissues in addition to activating immunoglobulin gene expression. It also modulates transcription transactivation by NR3C1, AR and PGR. Oct2 is B-cell specific, belongs to the POU transcription factor family class-2 subfamily and contains 1 homeobox domain.

Specifications

Antigen OCT2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene POU2F2
Gene Accession No. P09086, Q00196
Gene Alias Oct 2, OCT2, Octamer binding protein 2, OTF 2, OTF2, POU class 2 homeobox 2, POU2F2
Gene Symbols Pou2f2
Host Species Rabbit
Immunogen OCT2 Fusion Protein Ag0955
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18987, 5452
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.