Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980988
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0988 Supplier Bioss Supplier No. BS1077R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OCT-2 is a transcription factor belonging to the POU homeo-domain family that binds to the Ig gene octamer sites regulating B cell specific genes. OCT-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. OCT-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.

Specifications

Antigen OCT2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene POU2F2
Gene Accession No. P09086
Gene Alias 2-Oct; homeobox protein; Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; Oct2; Oct-2; Oct2a; Oct2b; Octamer-binding protein 2; Octamer-binding transcription factor 2; Otf2; OTF-2; POU class 2 homeobox 2; POU domain, class 2, transcription factor 2; POU2F2
Gene Symbols POU2F2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse OCT2 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5452
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.