Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OCT4 Antibody (CL7621), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32115825UL
Description
OCT4 Monoclonal antibody specifically detects OCT4 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
OCT4 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
CL7621 | |
Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein | |
This antibody was developed against a Synthetic Peptide of Q01860. Actual immunogen sequence is proprietary. | |
25 μg | |
Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators | |
5460 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction