Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP259164

Catalog No. NBP259164


Only null left
Add to Cart

Description

Description

44473 Polyclonal specifically detects 44473 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Chromatin Immunoprecipitation Sequencing.
Specifications

Specifications

OCT4
Polyclonal
Western Blot 1:1000, ELISA 1:1000-1:10000, Chromatin Immunoprecipitation (ChIP) 5 ug/IP, Chromatin Immunoprecipitation Sequencing
class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein
Rabbit
Affinity Purified
RUO
Primary
Human
IgG
Western Blot, ELISA, ChIP Assay
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
POU5F1
The exact immunogen is propietary information.
50 μg
Cellular Markers, Chromatin Research, Embryonic Stem Cell Markers, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators
5460
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only