Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OCT4 Antibody (OTI9B7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271225B
Description
44473 Monoclonal specifically detects 44473 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
OCT4 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
POU5F1 | |
Full length recombinant protein of human OCT4 (NP_002692) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI9B7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein | |
Mouse | |
Affinity Purified | |
Cellular Markers, Chromatin Research, Embryonic Stem Cell Markers, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators | |
5460.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction