Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OCT6 Polyclonal Antibody

OCT6 Polyclonal Antibody
Supplier: Thermo Scientific PA536258
Description
OCT6 Polyclonal Antibody for Western Blot

Specifications
OCT6 | |
Polyclonal | |
Unconjugated | |
POU3F1 | |
6-Oct; LOW QUALITY PROTEIN: POU domain, class 3, transcription factor 1; Oct6; Oct-6; octamer binding factor oct6; octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; Otf6; OTF-6; POU class 3 homeobox 1; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1; Scip; suppressed cAMP-inducible POU; Test1; Testes-1; Tst1; Tst-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18991, 192110, 5453 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.2 | |
P20267, P21952, Q03052 | |
POU3F1 | |
Synthetic peptide corresponding to amino acids 300-350 of human Oct-6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction