Learn More
Invitrogen™ OCT6 Polyclonal Antibody

Description
Immunogen sequence: MATTAQYLPR GPGGGAGGTG PLMHPDAAAA AAAAAAAERL HAGAAYREVQ KLMHHEWLGA GAGHPVGLAH PQWLPTGGGG GGDWAGGPHL EHGKAGGGGT GRADDGGGGG GFHARLVHQG AAHAGAAWAQ GSTAHHLGPA MSPSPGASGG HQPQPLGLYA QAAYPGGGGG GLAGMLAAGG GGAGPGLHHA LHEDGHEAQL EPSPPPHLGA HGHAHGHAHA GGLHAAAAHL HPGAGGGGSS VGEHSDEDAP SSDDLEQFAK.
Specifications
Specifications
| Antigen | OCT6 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.98 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | POU3F1 |
| Gene Accession No. | P20267, P21952 |
| Gene Alias | 6-Oct; LOW QUALITY PROTEIN: POU domain, class 3, transcription factor 1; Oct6; Oct-6; octamer binding factor oct6; octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; Otf6; OTF-6; POU class 3 homeobox 1; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1; Scip; suppressed cAMP-inducible POU; Test1; Testes-1; Tst1; Tst-1 |
| Gene Symbols | POU3F1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.