Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OCT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111427
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111427 100 μL
PIPA5111714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111427 Supplier Invitrogen™ Supplier No. PA5111427
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AERLHAGAAY REVQKLMHHE WLGAGAGHPV GLAHPQWLPT GGGGGGDWAG GPHLEHGKAG Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 33%.

POU3F1 is a member of the POU domain family of proteins, and regulates events during neurogenesis and myelination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCT6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POU3F1
Gene Accession No. Q03052
Gene Alias 6-Oct; LOW QUALITY PROTEIN: POU domain, class 3, transcription factor 1; Oct6; Oct-6; octamer binding factor oct6; octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; Otf6; OTF-6; POU class 3 homeobox 1; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1; Scip; suppressed cAMP-inducible POU; Test1; Testes-1; Tst1; Tst-1
Gene Symbols POU3F1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human OCT6. Recombinant protein control fragment (Product #RP-106674).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5453
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.