Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OCT6 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537882
Description
This antibody is predicted to react with mouse, rat, dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
POU3F1 is a member of the POU domain family of proteins, and regulates events during neurogenesis and myelination.
Specifications
OCT6 | |
Polyclonal | |
Unconjugated | |
POU3F1 | |
6-Oct; LOW QUALITY PROTEIN: POU domain, class 3, transcription factor 1; Oct6; Oct-6; octamer binding factor oct6; octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; Otf6; OTF-6; POU class 3 homeobox 1; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1; Scip; suppressed cAMP-inducible POU; Test1; Testes-1; Tst1; Tst-1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
5453 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q03052 | |
POU3F1 | |
Peptide with sequence C-HEDGHEAQLEPSP, from the internal region of POU3F1 (aa192-204). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction