Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentadecane, 99%

Catalog No. AC129820010
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass bottle

CAS: 629-62-9 | C15H32 | 212.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-62-9
Molecular Formula C15H32
Molecular Weight (g/mol) 212.42
MDL Number MFCD00008990
InChI Key YCOZIPAWZNQLMR-UHFFFAOYSA-N
Synonym n-pentadecane, pentadekan, unii-16h6k2s8m2, ch3-ch2 13-ch3, mys, pentadecane, analytical standard, pentadecan, medicinal plant, pentadecyl group, pentadecane, n
PubChem CID 12391
ChEBI CHEBI:28897
IUPAC Name pentadecane
SMILES CCCCCCCCCCCCCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 9.9°C
Density 0.7730g/mL
Boiling Point 270.0°C
Flash Point 132°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4307 to 1.4327
Linear Formula CH3(CH2)13CH3
Quantity 1 L
Beilstein 01, 172
Specific Gravity 0.773
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 212.42
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Pentadecane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-098-1
RTECSNumber RZ1800000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.