Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecane, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187868 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 g
5 g
Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 455°F
CAS 593-45-3
Vapor Pressure 1 mmHg (119°C)
Vapor Density 8.8 (vs. air)
Density 0.777 g/mL (at 25°C)
Flash Point 166°C
Physical Form Neat
Linear Formula CH3(CH2)16 CH3
MDL Number MFCD00009007
Melting Point 26°C to 29°C; 28°C to 30°C
Boiling Point 317°C
Type Analytical Standard
Quantity 1 g
Formula Weight 254.49
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.