Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecanedioic Acid 98.0+%, TCI America™
SDP

Catalog No. O022225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O022225G 25 g
O02221G 1 g
O02225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O022225G Supplier TCI America Supplier No. O022225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 871-70-5
Molecular Formula C18H34O4
Molecular Weight (g/mol) 314.466
MDL Number MFCD00142369
InChI Key BNJOQKFENDDGSC-UHFFFAOYSA-N
Synonym 1,16-hexadecanedicarboxylic acid, 1,18-octadecanedioic acid, unii-rsz6pq0qqj, octadecane-1,18-dioic acid, rsz6pq0qqj, hexadecanedicarboxylic acid, 1,18-octadecadioic acid, octadecanedioate, 1,18-octadecanedioate, octadecandis
PubChem CID 70095
IUPAC Name octadecanedioic acid
SMILES C(CCCCCCCCC(=O)O)CCCCCCCC(=O)O

Specifications

Melting Point 125°C
Color White
Quantity 25 g
Formula Weight 314.47
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Octadecanedioic Acid
EINECSNumber (2)-4022
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.