Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octadecylsuccinic Anhydride 95.0+%, TCI America™
SDP

Catalog No. O0018250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0018250G 250 g
O001825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0018250G Supplier TCI America Supplier No. O0018250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 47458-32-2
Molecular Formula C22H40O3
Molecular Weight (g/mol) 352.559
MDL Number MFCD00014551
InChI Key ZJFCVUTYZHUNSW-UHFFFAOYSA-N
Synonym octadecylsuccinic anhydride, n-octadecylsuccinic anhydride, octadecyl succinic anhydride, 2,5-furandione, dihydro-3-octadecyl, 3-octadecyldihydrofuran-2,5-dione, 2-octadecylsuccinic anhydride, dihydro-3-octadecylfuran-2,5-dione, stearylsuccinic anhydride, acmc-1an3y, 2, dihydro-3-octadecyl
PubChem CID 96562
IUPAC Name 3-octadecyloxolane-2,5-dione
SMILES CCCCCCCCCCCCCCCCCCC1CC(=O)OC1=O

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 250 g
Physical Form Crystal-Powder at 20°C
Formula Weight 352.56
Percent Purity ≥95.0% (T)
Chemical Name or Material Octadecylsuccinic Anhydride
EINECSNumber (2)-0851&(2)-1134
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.