Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octafluoronaphthalene, 96%

Catalog No. p-7025954
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 313-72-4 | C10F8 | 272.097 g/mol

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water. Solubility in toluene, almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313-72-4
Molecular Formula C10F8
Molecular Weight (g/mol) 272.097
MDL Number MFCD00014307
InChI Key JDCMOHAFGDQQJX-UHFFFAOYSA-N
Synonym octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion
PubChem CID 67564
IUPAC Name 1,2,3,4,5,6,7,8-octafluoronaphthalene
SMILES C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F

Specifications

Melting Point 84°C to 89°C
Boiling Point 209°C
Quantity 1 g
Beilstein 1915630
Solubility Information Insoluble in water. Solubility in toluene,almost transparent.
Formula Weight 272.1
Percent Purity 96%
Chemical Name or Material Octafluoronaphthalene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-239-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.