Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octafluorotoluene, 98+%

Catalog No. p-7081431
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 434-64-0 | C7F8 | 236.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-64-0
Molecular Formula C7F8
Molecular Weight (g/mol) 236.06
MDL Number MFCD00000375
InChI Key USPWUOFNOTUBAD-UHFFFAOYSA-N
Synonym octafluorotoluene, perfluorotoluene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl benzene, toluene, octafluoro, benzene, pentafluoro trifluoromethyl, trifluoromethyl pentafluorobenzene, pentafluorobenzotrifluoride, 1,2,3,4,5-pentafluoro-6-trifluoromethyl-benzene, benzene, 1,2,3,4,5-pentafluoro-6-trifluoromethyl, acmc-209juh
PubChem CID 9906
IUPAC Name 1,2,3,4,5-pentafluoro-6-(trifluoromethyl)benzene
SMILES FC1=C(F)C(F)=C(C(F)=C1F)C(F)(F)F

Specifications

Melting Point -66°C
Density 1.668
Boiling Point 103°C to 105°C
Flash Point 20°C (68°F)
Refractive Index 1.368
Quantity 5 g
UN Number UN1992
Beilstein 2057072
Formula Weight 236.06
Percent Purity ≥98%
Chemical Name or Material Octafluorotoluene
Show More Show Less
Hazard Category H225-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H225-H301-H331-H315-H319-H335
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 207-104-7
RTECSNumber XT3530000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.